Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687588

Sigma-Aldrich

(S)-N-Boc-2-methylpyrrolidine

95%

Synonym(s):

(S)-2-Methylpyrrolidine-1-carboxylic acid-tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO2
CAS Number:
Molecular Weight:
185.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

form

liquid

refractive index

n20/D 1.444

density

0.962 g/mL at 25 °C

SMILES string

C[C@H]1CCCN1C(=O)OC(C)(C)C

InChI

1S/C10H19NO2/c1-8-6-5-7-11(8)9(12)13-10(2,3)4/h8H,5-7H2,1-4H3/t8-/m0/s1

InChI key

PPUYUEPZGGATCN-QMMMGPOBSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

180.0 °F - closed cup

Flash Point(C)

82.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service