Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687472

Sigma-Aldrich

Sodium palmitate-2,4,6,8,10,12,14,16-13C8, 2,2-d2

≥97 atom % D, ≥99 atom % 13C, ≥98% (CP)

Synonym(s):

Hexadecanoic acid-2,4,6,8,10,12,14,16-13C8, 2,2-d2 sodium salt, Palmitic acid-2,4,6,8,10,12,14,16-13C8, 2,2-d2 sodium salt, Sodium hexadecanoate-2,4,6,8,10,12,14,16-13C8, 2,2-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(CH213CH2)6CH213CD2CO2Na
Molecular Weight:
288.36
UNSPSC Code:
12142201

isotopic purity

≥99 atom % 13C
≥97 atom % D

Assay

≥98% (CP)

form

flakes

mass shift

M+10

SMILES string

[13CH3]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13C]([2H])([2H])C(O[Na])=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

402.8 °F - closed cup

Flash Point(C)

206 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service