Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687456

Sigma-Aldrich

Methyl N-Boc-2-pyrrolecarboxylate

97%

Synonym(s):

1-(tert-Butyl) 2-methyl pyrrole-1,2-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO4
Molecular Weight:
225.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.4848

density

1.0466 g/mL at 25 °C

SMILES string

COC(=O)c1cccn1C(=O)OC(C)(C)C

InChI

1S/C11H15NO4/c1-11(2,3)16-10(14)12-7-5-6-8(12)9(13)15-4/h5-7H,1-4H3

InChI key

BLKOYCNYVYTNAG-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service