Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687251

Sigma-Aldrich

Tris[4-(5-dicyanomethylidenemethyl-2-thienyl)phenyl]amine

97%

Synonym(s):

TDCV-TPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H21N7S3
CAS Number:
Molecular Weight:
719.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

400 °C

fluorescence

λex 510 nm; λem 592 nm in chloroform

Orbital energy

HOMO 6 eV 
LUMO 4.2 eV 

OPV Device Performance

ITO/PEDOT:PSS/TDCV-TPA/Al

  • Short-circuit current density (Jsc): 1.7 mA/cm2
  • Open-circuit voltage (Voc): 0.76 V
  • Fill Factor (FF): 0.27
  • Power Conversion Efficiency (PCE): 0.4 %

ITO/PEDOT:PSS/TDCV-TPA/C60/LiF/AL
  • Short-circuit current density (Jsc): 4.59 mA/cm2
  • Open-circuit voltage (Voc): 1.15 V
  • Fill Factor (FF): 0.28
  • Power Conversion Efficiency (PCE): 1.85 %

ITO/PEDOT:PSS/TDCV-TPA/PC61BM/Al
  • Short-circuit current density (Jsc): 3.65 mA/cm2
  • Open-circuit voltage (Voc): 0.96 V
  • Fill Factor (FF): 0.29
  • Power Conversion Efficiency (PCE): 1.02 %

SMILES string

N#C\C(=C/c1ccc(s1)-c2ccc(cc2)N(c3ccc(cc3)-c4ccc(\C=C(/C#N)C#N)s4)c5ccc(cc5)-c6ccc(\C=C(/C#N)C#N)s6)C#N

InChI

1S/C42H21N7S3/c43-22-28(23-44)19-37-13-16-40(50-37)31-1-7-34(8-2-31)49(35-9-3-32(4-10-35)41-17-14-38(51-41)20-29(24-45)25-46)36-11-5-33(6-12-36)42-18-15-39(52-42)21-30(26-47)27-48/h1-21H

InChI key

KJHGHQMIPHPKIY-UHFFFAOYSA-N

Application

Isotropic organic semiconductor for OPV′s and OLED′s. TDCV-TPA is soluble in a large variety of organic solvents and can be used for the fabrication of heterojunction solar cells with high open-circuit voltage (1.15V), efficiency close to 2% and longer ambient condition lifetimes than cells based on poly(alkylthiophenes). The compound absorbs at 509nm (in CH2Cl2 solution) and 538nm as thin film. It can be used as high spectral purity luminophore in LED′s, emitting red light at 658nm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cravino, A.; Leriche, P.; Aleveque, O.; Roncali, J.
Advanced Materials, 18, 3033-3033 (2006)
Sophie Roquet et al.
Journal of the American Chemical Society, 128(10), 3459-3466 (2006-03-09)
Star-shaped molecules based on a triphenylamine core derivatized with various combinations of thienylenevinylene conjugated branches and electron-withdrawing indanedione or dicyanovinyl groups have been synthesized. UV-vis absorption and fluorescence emission data show that the introduction of the electron-acceptor groups induces an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service