Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686557

Sigma-Aldrich

2-Methylpropane-1-13C

99 atom % 13C

Synonym(s):

Isobutane-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH13CH3
Molecular Weight:
59.11
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

form

gas

bp

-12 °C (lit.)

mp

-160 °C (lit.)

mass shift

M+1

SMILES string

CC(C)[13CH3]

InChI

1S/C4H10/c1-4(2)3/h4H,1-3H3/i1+1

InChI key

NNPPMTNAJDCUHE-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-117.4 °F - closed cup

Flash Point(C)

-83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service