Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686077

Sigma-Aldrich

Scandium(III) chloride

hydrogen-storage grade, 99.9%

Synonym(s):

Scandium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ScCl3
CAS Number:
Molecular Weight:
151.31
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
grade:
hydrogen-storage grade
form:
powder

grade

hydrogen-storage grade

Quality Level

Assay

99.9%

form

powder

mp

960 °C (lit.)

density

2.39 g/mL at 25 °C (lit.)

SMILES string

Cl[Sc](Cl)Cl

InChI

1S/3ClH.Sc/h3*1H;/q;;;+3/p-3

InChI key

DVMZCYSFPFUKKE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Analysis Note

XRD plots and metal purity data are available upon request.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service