Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

685410

Sigma-Aldrich

Methyl 1-acetyl-1H-pyrrole-3-carboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

powder

mp

91-97 °C

functional group

ester

SMILES string

COC(=O)c1ccn(c1)C(C)=O

InChI

1S/C8H9NO3/c1-6(10)9-4-3-7(5-9)8(11)12-2/h3-5H,1-2H3

InChI key

XNEPOBYEFLQFMM-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service