Skip to Content
MilliporeSigma
All Photos(1)

Documents

684953

Sigma-Aldrich

4-Fluoro-2-methylbenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.5260

density

1.144 g/mL at 25 °C

SMILES string

[H]C(=O)c1ccc(F)cc1C

InChI

1S/C8H7FO/c1-6-4-8(9)3-2-7(6)5-10/h2-5H,1H3

InChI key

ADCFIKGEGWFWEA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

180.0 °F - closed cup

Flash Point(C)

82.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service