Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

684473

Sigma-Aldrich

[6,6]-Phenyl C85 butyric acid methyl ester, mixture of isomers

≥99%

Synonym(s):

Methyl 3′-phenyl-3′H-cyclopropa[18,7][5,6]fullerene-C84-D2d-3′-butanoate, [84]PCBM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H14O2
CAS Number:
Molecular Weight:
1199.14
UNSPSC Code:
12352103

Assay

≥99%

solubility

chlorobenzene: soluble
organic solvents: soluble
toluene: soluble

SMILES string

COC(=O)CCCC1(C2c3cc4cc5cc6C7CC8CC9CCC%10CCC%11CC%12CCC12c%13c%12c%14C%11c%15c%10c%16c9c%17C8C7=C%18c%17c%19c%16c%15c%20c%14c%21c%13c3c%22c4c%23c5c6c%18c%24c%19c%20c%21c%22c%23%24)c%25ccccc%25

Application

Soluble n-channel organic semiconductor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kooistra, F.; Mihailetchi, V. et al.
Chemistry of Materials, 18, 3068-3068 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service