Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68416

Sigma-Aldrich

(R)-(+)-1-Benzylpyrrolidine-2-methanol

≥95.0% (GC)

Synonym(s):

(R)-(+)-1-Benzyl-2-(hydroxymethyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥95.0% (GC)

optical activity

[α]/D +74±2°, neat

refractive index

n20/D 1.541

SMILES string

OC[C@H]1CCCN1Cc2ccccc2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service