Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

683965

Sigma-Aldrich

2,4-Bis(trifluoromethyl)phenylboronic acid

≥95%

Synonym(s):

(2,4-Ditrifluoromethylphenyl)boronic acid, 2,4-Bis(trifluoromethyl)benzeneboronic acid, 2,4-Di(trifluoromethyl)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5BF6O2
CAS Number:
Molecular Weight:
257.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

crystals

mp

110-117 °C

functional group

fluoro

SMILES string

OB(O)c1ccc(cc1C(F)(F)F)C(F)(F)F

InChI

1S/C8H5BF6O2/c10-7(11,12)4-1-2-6(9(16)17)5(3-4)8(13,14)15/h1-3,16-17H

InChI key

WLYPBMBWKYALCG-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Pyrrolo[2,3-d]pyrimidin-4-ones as corticotropin-releasing factor 1 receptor antagonists with a carbonyl-based hydrogen bonding acceptor
  • Biaryl pyrazole carboxamides as sodium channel blockers for treatment of neuropathic pain
  • Biphenylketones as dual modulators of inflammation and bone loss
  • (Biphenylyl)pyrazolecarboxamides and their voltage-gated sodium channel Nav1.7 blocking structure-activity relationship
  • N-hydroxyalkyl biphenylsulfonamides as inhibitors of bone resorption
  • 1,3,6,8a-tetraazaacenaphthylene derivatives and study of their activity as corticotropin-releasing factor-1 antagonists
  • Blue luminescent iridium phenylpyridine complexes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service