Skip to Content
MilliporeSigma
All Photos(1)

Documents

682713

Sigma-Aldrich

3-Nitrophenylboronic acid L-tartaric acid ester solution

1.0 M in THF

Synonym(s):

2-(3-Nitrophenyl)-1,3,2-dioxaborolane-4R,5R-dicarboxylic acid, L-TarB-NO2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BNO8
CAS Number:
Molecular Weight:
280.98
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

liquid

optical activity

[α]22/D +9.0°, neat

concentration

1.0 M in THF

refractive index

n20/D 1.4508

density

1.0525 g/mL at 25 °C

SMILES string

OC(=O)[C@@H]1OB(O[C@H]1C(O)=O)c2cccc(c2)[N+]([O-])=O

InChI

1S/C10H8BNO8/c13-9(14)7-8(10(15)16)20-11(19-7)5-2-1-3-6(4-5)12(17)18/h1-4,7-8H,(H,13,14)(H,15,16)/t7-,8-/m1/s1

InChI key

DKYPDMHYDHHGRL-HTQZYQBOSA-N

Application

In conjunction with NaBH4, this Lewis acid reduces prochiral alkyl aryl ketones with excellent enantioselectivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.0 °F

Flash Point(C)

-17.78 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim, J.; Singaram, B.
Tetrahedron Letters, 47, 3901-3901 (2006)
Kim, J. et al.
Organic Process Research & Development, 10, 949-949 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service