Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682438

Sigma-Aldrich

2-Nitro-6-(trifluoromethyl)aniline

97%

Synonym(s):

α,α,α-Trifluoro-6-nitro-ortho-toluidine, 2-Amino-3-nitrobenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5F3N2O2
CAS Number:
Molecular Weight:
206.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

48-52 °C

SMILES string

Nc1c(cccc1C(F)(F)F)[N+]([O-])=O

InChI

1S/C7H5F3N2O2/c8-7(9,10)4-2-1-3-5(6(4)11)12(13)14/h1-3H,11H2

InChI key

MFLJFFUIURSSKM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service