Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682268

Sigma-Aldrich

L-Aspartic acid-13C4,15N,2,3,3-d3

98 atom % D, 99 atom % 13C, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4D3H415NO4
Molecular Weight:
141.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N
98 atom % D

mass shift

M+8

SMILES string

[2H][13C]([2H])([13C](O)=O)[13C@]([2H])([15NH2])[13C](O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1/i1+1D2,2+1D,3+1,4+1,5+1

InChI key

CKLJMWTZIZZHCS-ULNHBARWSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service