682063
1-Methyl-1H-pyrazole-4-carboxylic acid
97%
Synonym(s):
1-Methyl-4-pyrazolecarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
203-208 °C
SMILES string
Cn1cc(cn1)C(O)=O
InChI
1S/C5H6N2O2/c1-7-3-4(2-6-7)5(8)9/h2-3H,1H3,(H,8,9)
InChI key
UPPPWUOZCSMDTR-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service