Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682055

Sigma-Aldrich

5-Hydroxy-1-methyl-1H-pyrazole

95%

Synonym(s):

1-Methyl-1H-pyrazol-5-ol, 1-Methyl-5-hydroxy-1H-pyrazole, 1-Methyl-5-hydroxypyrazole, 1-Methylpyrazol-5-ol, 2-Methyl-2H-pyrazol-3-ol, 5-Hydroxy-1-methylpyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

110-114 °C

SMILES string

Cn1nccc1O

InChI

1S/C4H6N2O/c1-6-4(7)2-3-5-6/h2-3,7H,1H3

InChI key

CMXOTACIOGGSNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service