Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680664

Sigma-Aldrich

3-Bromo-5-formyl-2-methoxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8BBrO4
CAS Number:
Molecular Weight:
258.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

200-211 °C

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

COc1c(Br)cc(C=O)cc1B(O)O

InChI

1S/C8H8BBrO4/c1-14-8-6(9(12)13)2-5(4-11)3-7(8)10/h2-4,12-13H,1H3

InChI key

UUPRSFVSYMWVRC-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service