Skip to Content
MilliporeSigma
All Photos(2)

Documents

680613

Sigma-Aldrich

N-Boc-5-methoxyindole

97%

Synonym(s):

5-Methoxyindole-1-carboxylic acid tert-butyl ester, N-(tert-Butoxycarbonyl)-5-methoxyindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

75-79 °C

SMILES string

COc1ccc2n(ccc2c1)C(=O)OC(C)(C)C

InChI

1S/C14H17NO3/c1-14(2,3)18-13(16)15-8-7-10-9-11(17-4)5-6-12(10)15/h5-9H,1-4H3

InChI key

LPSLGTZFBDZNEK-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Asymmetric intramolecular Friedel-Crafts alkylation reaction
  • Palladium-catalyzed carboaminoxylations with arylboronic acids and TEMPO catalyst
  • Fluorescent pyrimidopyrimidoindole nucleosides via Suzuki-Miyaura coupling reaction
  • Novel conformationally restricted β- and γ-amino acids
  • 2-(Indolyl) borates, silanes, and silanols
  • DNA minor groove alkylating agents as stable amine-based prodrugs designed for tumor-specific release

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manabu Nakazono et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 27-34 (2014-05-13)
Various styrylbenzene compounds were synthesized and evaluated as mainly Aβ amyloid sensors. These compounds, however, cannot be used for detecting amyloid deposition in peripheral nerves because of the inherent sensitivity of the compounds. These compounds often generate false positives especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service