Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680575

Sigma-Aldrich

3-Vinylphenylboronic acid

≥95%

Synonym(s):

(3-Ethenylphenyl)boronic acid, m-Vinylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BO2
CAS Number:
Molecular Weight:
147.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

powder or crystals

mp

141-147 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cccc(C=C)c1

InChI

1S/C8H9BO2/c1-2-7-4-3-5-8(6-7)9(10)11/h2-6,10-11H,1H2

InChI key

SYBQEKBVWDPVJM-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant involved in:
  • Suzuki-Miyaura reactions
  • Molecular imprinting of fructose and pinacol
  • Synthesis of arylmercapturic acids via Chen-Lam-Evans arylation
  • Three-component cross-coupling yeilding diarylketones
  • Generation of acyl anion equivalents

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengzhe Guo et al.
Journal of separation science, 39(17), 3428-3435 (2016-07-13)
Maltose, a common auxiliary material of pharmaceutical preparation, may disturb the analysis of total amino acids in sepia capsule by aldolization. Therefore, it is necessary to remove the maltose through a convenient method. In this work, a phenylboronic acid modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service