Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

679917

Sigma-Aldrich

4,4′-Dibromotriphenylamine

96%

Synonym(s):

N,N-Bis(4-bromophenyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13Br2N
CAS Number:
Molecular Weight:
403.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

SMILES string

Brc1ccc(cc1)N(c2ccccc2)c3ccc(Br)cc3

InChI

1S/C18H13Br2N/c19-14-6-10-17(11-7-14)21(16-4-2-1-3-5-16)18-12-8-15(20)9-13-18/h1-13H

InChI key

KIGVOJUDEQXKII-UHFFFAOYSA-N

Application

Hole transport material and building block for hole transport materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polytriarylamine Semiconductors

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service