Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679526

Sigma-Aldrich

QuadraSil® MP

extent of labeling: 1.0-1.5 mmol/g loading

Synonym(s):

QuadraSil® Mercaptopropyl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

extent of labeling

1.0-1.5 mmol/g loading

Application

  • Metal scavenger
  • Used for catalytic Suzuki-Miyaura reaction inhibition by entraining catalytic palladium nanoparticles
Metal scavenger: Pd (with or without phosphines), Rh, Cu, Ru, Pt, Pb, Ag, Hg

Legal Information

QuadraSil is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service