Skip to Content
MilliporeSigma
All Photos(1)

Documents

679496

Sigma-Aldrich

QuadraSil® TA

extent of labeling: 0.5-1.5 mmol/g loading

Synonym(s):

QuadraSil® Triamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

extent of labeling

0.5-1.5 mmol/g loading

Application

Metal scavenger: Au, Cd, Co, Cu, Fe, Pd, Pt, Rh, Ru, V, Zn, Pb

Legal Information

QuadraSil is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junichi Nakajima et al.
Talanta, 79(4), 1050-1054 (2009-07-21)
A flow injection analysis (FIA) method using on-line separation and preconcentration with a novel metal scavenger beads, QuadraSil TA, has been developed for the ICP-OES determination of traces of palladium. QuadraSil TA contains diethylenetriamine as a functional group on spherical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service