Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679224

Sigma-Aldrich

Butyltrichlorosilane

≥98%, deposition grade

Synonym(s):

Trichlorobutylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SiCl3
CAS Number:
Molecular Weight:
191.56
Beilstein:
1736032
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥98%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

149 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CCCC[Si](Cl)(Cl)Cl

InChI

1S/C4H9Cl3Si/c1-2-3-4-8(5,6)7/h2-4H2,1H3

InChI key

FQEKAFQSVPLXON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Silica's versatility spans various industries, including biomedical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service