Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

678597

Sigma-Aldrich

2-Methylfuran-3-carbonyl chloride

97%

Synonym(s):

2-Methyl-3-furoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClO2
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5052

bp

59-61 °C/12 mmHg

density

1.239 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1occc1C(Cl)=O

InChI

1S/C6H5ClO2/c1-4-5(6(7)8)2-3-9-4/h2-3H,1H3

InChI key

AFFZVBHMMXZNKE-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

169.0 °F - closed cup

Flash Point(C)

76.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service