Skip to Content
MilliporeSigma
All Photos(1)

Documents

678449

Sigma-Aldrich

4-[(4-Methyl-1-piperazinyl)methyl]benzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21N3
CAS Number:
Molecular Weight:
219.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.547

density

1.0154 g/mL at 25 °C

SMILES string

CN1CCN(CC1)Cc2ccc(CN)cc2

InChI

1S/C13H21N3/c1-15-6-8-16(9-7-15)11-13-4-2-12(10-14)3-5-13/h2-5H,6-11,14H2,1H3

InChI key

HILINUGDTTXRNZ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service