Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

678317

Sigma-Aldrich

Tri-n-butyllithium magnesate

0.7 M in diethyl ether/hexanes

Synonym(s):

Lithium tributylmagnesate in hexanes/ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27LiMg
CAS Number:
Molecular Weight:
202.59
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.7 M in diethyl ether/hexanes

density

0.769 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].CCCC[Mg-](CCCC)CCCC

InChI

1S/3C4H9.Li.Mg/c3*1-3-4-2;;/h3*1,3-4H2,2H3;;/q;;;+1;-1

InChI key

MYNWCAJLXIFFBO-UHFFFAOYSA-N

General description

Tri-n-butyllithium magnesate is a base and deprotonating agent used in the deprotonation of furans, fluoro aromatics, thiophenes, oxazoles and benzoxazoles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-13.0 °F

Flash Point(C)

-25 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deprotonation of benzoxazole and oxazole using lithium magnesates
Bayh O, et al.
The Journal of Organic Chemistry, 70(13), 5190-5196 (2005)
Deprotonation of thiophenes using lithium magnesates
Bayh O, et al.
Tetrahedron, 61(20), 4779-4784 (2005)
Deprotonation of furans using lithium magnesates
Mongin F, et al.
Tetrahedron Letters, 46(46), 7989-7992 (2005)
Deprotonation of fluoro aromatics using lithium magnesates.
Awad H, et al.
Tetrahedron Letters, 45(36), 6697-6701 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service