Skip to Content
MilliporeSigma
All Photos(1)

Documents

678066

Sigma-Aldrich

Poly{[2,5-bis(2-(N,N-diethylamino)ethoxy)-1,4-phenylene]-alt-1,4-phenylene}

Synonym(s):

Poly{[2,5-bis(3-(N,N-diethylamino)-1-oxapropyl)-1,4-phenylene]-alt-1,4-phenylene}

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C24H34N2O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

λmax

340 nm

fluorescence

λem 401 nm in chloroform

Application

A neutral analogue of water-soluble quaternary ammonium functionalized poly(p-phenylene) (PPP) polyelectrolyte.

Soluble in organic solvents (THF, cholorform) and dilute aqueous acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.R. Reynolds
Macromolecules, 32, 3970-3970 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service