Skip to Content
MilliporeSigma
All Photos(3)

Documents

676624

Sigma-Aldrich

5-Methoxy-3-pyridineboronic acid pinacol ester

97%

Synonym(s):

(3-Methoxypyridin-5-yl)boronic acid pinacol ester, (5-Methoxypyridin-3-yl)boronic acid pinacol ester, 2-(5-Methoxypyridin-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine, 3-Methoxy-5-(pinacolboranato)pyridine, 5-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18BNO3
CAS Number:
Molecular Weight:
235.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

100-108 °C

SMILES string

COc1cncc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C12H18BNO3/c1-11(2)12(3,4)17-13(16-11)9-6-10(15-5)8-14-7-9/h6-8H,1-5H3

InChI key

HENXUFOAGXNWKH-UHFFFAOYSA-N

Application

5-Methoxy-3-pyridineboronic acid pinacol ester can be used:
  • To synthesize anthracene based bis-pyridine ligand (L), which is employed in the preparation of fluorescent M2L4 type capsules (M= Pt, Zn, Pd, Cu, Ni, Co, and Mn).
  • As a starting material in the synthesis of pyridylmethyl pyridine derivatives as potent 11β-hydroxylase (CPY11B1) inhibitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cushing?s syndrome: Development of highly potent and selective CYP11B1 inhibitors of the (pyridylmethyl) pyridine type
Emmerich J, et al.
Journal of Medicinal Chemistry, 56(15), 6022-6032 (2013)
Isostructural M2L4 molecular capsules with anthracene shells: synthesis, crystal structures, and fluorescent properties
Li Z, et al.
Chemistry?A European Journal , 18(27), 8358-8365 (2012)
Accelerated skin wound healing by selective 11β-hydroxylase (CYP11B1) inhibitors
Emmerich J, et al.
European Journal of Medicinal Chemistry, 143(27), 591-597 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service