Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676241

Sigma-Aldrich

Diiodo(p-cymene)ruthenium(II) dimer

Synonym(s):

Di-μ-iodo(p-cymene)iodoruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28I4Ru2
CAS Number:
Molecular Weight:
978.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mp

234-236 °C

SMILES string

I[Ru]1[I][Ru](I)[I]1.C[C]2[CH][CH][C]([CH][CH]2)C(C)C.C[C]3[CH][CH][C]([CH][CH]3)C(C)C

InChI

1S/2C10H14.2HI.2I.2Ru/c2*1-8(2)10-6-4-9(3)5-7-10;;;;;;/h2*4-8H,1-3H3;2*1H;;;;/q;;;;;;2*+1/p-2

InChI key

GMEPSZJJBHGIDB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst in:
  • Ruthenium-catalyzed hydrogenation
  • Preparation of Walphos-type ferrocenyl-aryl diphosphine ligands
  • Ruthenium-catalyzed synthesis reactions
  • Enantioselective hydrogenation of ketones and olefins

Legal Information

A product of Umicore

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service