Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

675849

Sigma-Aldrich

Hexa(ethylene glycol)mono-11-(acetylthio)undecyl ether

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H50O8S
CAS Number:
Molecular Weight:
510.72
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

refractive index

n20/D 1.4782

density

1.0474 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC(=O)SCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C25H50O8S/c1-25(27)34-24-10-8-6-4-2-3-5-7-9-12-28-14-16-30-18-20-32-22-23-33-21-19-31-17-15-29-13-11-26/h26H,2-24H2,1H3

InChI key

YQZGQQAPWBBCSS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Hydrolyzing agents deprotect protected thiol derivatives, obtaining unstable free thiols during complex compound synthesis for self-assembled monolayers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service