Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67510

Sigma-Aldrich

5-Methyl-1-hexene

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2CH=CH2
CAS Number:
Molecular Weight:
98.19
Beilstein:
1731846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
≥99.0%

Assay

≥99.0%

refractive index

n20/D 1.396

bp

84-85 °C (lit.)

density

0.691 g/mL at 20 °C (lit.)

SMILES string

CC(C)CCC=C

InChI

1S/C7H14/c1-4-5-6-7(2)3/h4,7H,1,5-6H2,2-3H3

InChI key

JIUFYGIESXPUPL-UHFFFAOYSA-N

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service