Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

673951

Sigma-Aldrich

Tricyclopentylphosphine tetrafluoroborate

greener alternative

95%

Synonym(s):

Cyp3HBF4, PCyp3HBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28BF4P
CAS Number:
Molecular Weight:
326.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

246-264 °C

functional group

phosphine

greener alternative category

SMILES string

F[B-](F)(F)F.C1CCC(C1)[PH+](C2CCCC2)C3CCCC3

InChI

1S/C15H27P.BF4/c1-2-8-13(7-1)16(14-9-3-4-10-14)15-11-5-6-12-15;2-1(3,4)5/h13-15H,1-12H2;/q;-1/p+1

InChI key

KTDMVANQORSVPI-UHFFFAOYSA-O

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for alkane C-H arylation from aryl chlorides
Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.

Amide Synthesis from Alcohols and Amines by the Extrusion of Dihydrogen
Used with Ru(cod)Cl dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Acetylene chemistry's versatile applications from organic synthesis to bioorganic chemistry demand efficient synthesis methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service