Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

673927

Sigma-Aldrich

Ethyl N-Boc-4-methylpiperidine-4-carboxylate

97%

Synonym(s):

1-tert-Butoxycarbonyl-4-methylpiperidine-4-carboxylic acid ethyl ester, 4-Methyl-1,4-piperidinedicarboxylic acid 1-(1,1-dimethylethyl) 4-ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO4
CAS Number:
Molecular Weight:
271.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4554

density

1.0134 g/mL at 25 °C

SMILES string

CCOC(=O)C1(C)CCN(CC1)C(=O)OC(C)(C)C

InChI

1S/C14H25NO4/c1-6-18-11(16)14(5)7-9-15(10-8-14)12(17)19-13(2,3)4/h6-10H2,1-5H3

InChI key

ZQZVWDXMUCTNRI-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Dipeptidyl peptidase-4 inhibitor ABT-279
  • Building blocks for piperazine-based CCR5 antagonists
Replacement of the ester function with an amino group provides a key building block for piperazine-based CCR5 antagonists.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Hua Jiang et al.
Bioorganic & medicinal chemistry letters, 14(14), 3675-3678 (2004-06-19)
4-Substituted-4-aminopiperidine is an interesting structural motif found in a number of bioactive compounds. An efficient and convenient method for the synthesis of 4-differently substituted-4-aminopiperidine derivatives was described, employing isonipecotate as a starting material and Curtius rearrangement as a key step.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service