Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

673056

Sigma-Aldrich

2-(4-Boc-piperazinyl)-2-phenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N2O4
CAS Number:
Molecular Weight:
320.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

200-204 °C

SMILES string

CC(C)(C)OC(=O)N1CCN(CC1)C(C(O)=O)c2ccccc2

InChI

1S/C17H24N2O4/c1-17(2,3)23-16(22)19-11-9-18(10-12-19)14(15(20)21)13-7-5-4-6-8-13/h4-8,14H,9-12H2,1-3H3,(H,20,21)

InChI key

QPEHPIVVAWESTM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service