Skip to Content
MilliporeSigma
All Photos(1)

Documents

672726

Sigma-Aldrich

2-Bromo-N-decyl-acetamide

97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24BrNO
CAS Number:
Molecular Weight:
278.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97% (GC)

form

solid

mp

41-45 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCNC(=O)CBr

InChI

1S/C12H24BrNO/c1-2-3-4-5-6-7-8-9-10-14-12(15)11-13/h2-11H2,1H3,(H,14,15)

InChI key

RZTGBAJAVREPOB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Y Lee et al.
The Journal of biological chemistry, 255(13), 6121-6127 (1980-07-10)
Human serum albumin (HSA) is found to contain one primary binding site for medium chain fatty acids which is competitive with the binding of N-acetyl-L-tryptophan. The association constant is higher with nondefatted HSA than with HSA defatted by either heptane-acetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service