Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672548

Sigma-Aldrich

6-Chloro-2-methoxy-9-phenoxyacridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClNO2
CAS Number:
Molecular Weight:
335.78
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97.5-102.5% (T)
98%

mp

153-156 °C

SMILES string

COc1ccc2nc3cc(Cl)ccc3c(Oc4ccccc4)c2c1

InChI

1S/C20H14ClNO2/c1-23-15-8-10-18-17(12-15)20(24-14-5-3-2-4-6-14)16-9-7-13(21)11-19(16)22-18/h2-12H,1H3

InChI key

CKWITQOQGJPKOE-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service