Skip to Content
MilliporeSigma
All Photos(1)

Documents

672505

Sigma-Aldrich

N-Ethylpyridinium bromide-d10

for NMR spectroscopy, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D10BrN
CAS Number:
Molecular Weight:
198.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

for NMR spectroscopy

isotopic purity

≥99 atom % D

Assay

99%

composition

carbon, 41.80-43.08%
nitrogen, 6.96-7.18%

mass shift

M+10

storage temp.

2-8°C

SMILES string

[Br-].[2H]c1c([2H])c([2H])[n+](c([2H])c1[2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C7H10N.BrH/c1-2-8-6-4-3-5-7-8;/h3-7H,2H2,1H3;1H/q+1;/p-1/i1D3,2D2,3D,4D,5D,6D,7D;

InChI key

ABFDKXBSQCTIKH-BCYFSIEASA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service