Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672378

Sigma-Aldrich

2-Chloro-4,6-bis[3-(perfluorohexyl)propyloxy]-1,3,5-triazine

98% (H-NMR)

Synonym(s):

2-Chloro-4,6-bis(4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononyloxy)-1,3,5-triazine, F26 CDMT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H12ClF26N3O2
CAS Number:
Molecular Weight:
867.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98% (H-NMR)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCOc1nc(Cl)nc(OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)n1

InChI

1S/C21H12ClF26N3O2/c22-7-49-8(52-5-1-3-10(23,24)12(27,28)14(31,32)16(35,36)18(39,40)20(43,44)45)51-9(50-7)53-6-2-4-11(25,26)13(29,30)15(33,34)17(37,38)19(41,42)21(46,47)48/h1-6H2

InChI key

CBCNBGCPSUHKHN-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service