Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672122

Sigma-Aldrich

2-(Dicyclohexylphosphino)-1-(2,4,6-trimethyl-phenyl)-1H-imidazole

95%

Synonym(s):

2-(Dicyclohexylphosphino)-1-mesityl-1H-imidazole, cataCXium® PICy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H35N2P
CAS Number:
Molecular Weight:
382.52
Beilstein:
9956113
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

Cc1cc(C)c(c(C)c1)-n2ccnc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C24H35N2P/c1-18-16-19(2)23(20(3)17-18)26-15-14-25-24(26)27(21-10-6-4-7-11-21)22-12-8-5-9-13-22/h14-17,21-22H,4-13H2,1-3H3

InChI key

ZRVANNJGPCSNAH-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Application

Catalyst for coupling reactions of aryl chlorides

Legal Information

WO 2004/101581
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service