Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

671843

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoroacetate

for catalysis, ≥97.0% (HPLC)

Synonym(s):

[EMIM][TFA]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11F3N2O2
CAS Number:
Molecular Weight:
224.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (HPLC)

impurities

≤500 ppm water

anion traces

chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

[O-]C(=O)C(F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2HF3O2/c1-3-8-5-4-7(2)6-8;3-2(4,5)1(6)7/h4-6H,3H2,1-2H3;(H,6,7)/q+1;/p-1

InChI key

JOKVYNJKBRLDAT-UHFFFAOYSA-M

General description

1-Ethyl-3-methylimidazolium trifluoroacetate is a non-volatile ionic liquid. The activity coefficient values at infinite dilution for different organic solutes in [EMIM][TFA] indicates that it can be considered as a solvent for the separation of aliphatic hydrocarbons from aromatic hydrocarbon.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activity coefficients at infinite dilution measurements for organic solutes and water in the ionic liquid 1-ethyl-3-methylimidazolium trifluoroacetate.
Domanska U and Marciniak A.
The Journal of Physical Chemistry B, 111(41), 11984-11988 (2007)
Nan Shi et al.
Bioresource technology, 283, 174-183 (2019-03-25)
Lignocellulose was oxidatively decomposed in a newly developed polyoxometalates-imidazolium ionic liquid mixture. Aromatic compounds covering acids, esters, ketones, aldehydes, and phenols were selectively produced under various conditions. 4-Hydroxylbenzoic acid was dominatingly yielded under low temperature and high oxidant concentration. Phenolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service