Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

671436

Sigma-Aldrich

(1S,2S)-trans-N-Boc-2-aminocyclopentanol

99%

Synonym(s):

((1S,2S)-2-Hydroxycyclopentyl)carbamic acid tert-butyl ester, tert-Butyl N-((2S,1S)-2-hydroxycyclopentyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO3
CAS Number:
Molecular Weight:
201.26
Beilstein:
5810036
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (GC)
99%

optical purity

enantiomeric ratio: ≥95:5%

SMILES string

CC(C)(C)OC(=O)N[C@H]1CCC[C@@H]1O

InChI

1S/C10H19NO3/c1-10(2,3)14-9(13)11-7-5-4-6-8(7)12/h7-8,12H,4-6H2,1-3H3,(H,11,13)/t7-,8-/m0/s1

InChI key

CGZQRJSADXRRKN-YUMQZZPRSA-N

Application

Reactant for:
  • Asymmetric synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors via stereoselective thioesterification, thioetherification, hydrolysis, heterocyclization and amination

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service