Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

671371

Sigma-Aldrich

(3αR,7αR)-1,3-Dimethyl-2-(1-piperidinyl)octahydro-1H-1,3,2-benzodiazaphosphole 2-oxide

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26N3OP
CAS Number:
Molecular Weight:
271.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥98.0% (GC)

optical activity

[α]/D -90.5±3°, c = 1.15 in chloroform

mp

108-112 °C

SMILES string

CN1[C@@H]2CCCC[C@H]2N(C)P1(=O)N3CCCCC3

InChI

1S/C13H26N3OP/c1-14-12-8-4-5-9-13(12)15(2)18(14,17)16-10-6-3-7-11-16/h12-13H,3-11H2,1-2H3/t12-,13-/m1/s1

InChI key

ILDAVWNADMBKGB-CHWSQXEVSA-N

Application

Organocatalyst: Asymmetric Allylation of Aldehydes with Chiral Lewis Bases, Review

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buono, G., et al.
Synlett, 377-388 (1999)
Denmark, S. E.. et al
The Journal of Organic Chemistry, 59, 6161-6163 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service