Skip to Content
MilliporeSigma
All Photos(2)

Documents

670731

Sigma-Aldrich

(S)-α,α-Bis(3,5-dimethylphenyl)-2-pyrrolidinemethanol

≥99% (HPLC)

Synonym(s):

(S)-(−)-2-[Bis(3,5-dimethylphenyl)hydroxymethyl]pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO
CAS Number:
Molecular Weight:
309.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99% (HPLC)

optical purity

enantiomeric ratio: 97.5% (HPLC)

mp

98-101 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)cc(c1)C(O)([C@@H]2CCCN2)c3cc(C)cc(C)c3

InChI

1S/C21H27NO/c1-14-8-15(2)11-18(10-14)21(23,20-6-5-7-22-20)19-12-16(3)9-17(4)13-19/h8-13,20,22-23H,5-7H2,1-4H3/t20-/m0/s1

InChI key

MXIOBZPVLNQGIU-FQEVSTJZSA-N

Application

(S)-α,α-Bis(3,5-dimethylphenyl)-2-pyrrolidinemethanol can be used as:
  • A chiral organocatalyst in the preparation of α-[(phenyl)methyl]thio]alkanal derivatives using aldehydes as starting materials and [(phenyl)methyl]thio]triazole as a sulfur electrophile.
  • A catalyst in the preparation of chiral bipyrazolidin-3-one derivatives by cycloadditions of azomethine imines with α,β-unsaturated aldehydes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diarylprolinol silyl ether salts as new, efficient, water-soluble, and recyclable organocatalysts for the asymmetric Michael addition on water
Zheng Z, et al.
Journal of the American Chemical Society, 132(1), 50-51 (2010)
Enantioselective organocatalyzed alpha sulfenylation of aldehydes.
Mauro Marigo et al.
Angewandte Chemie (International ed. in English), 44(5), 794-797 (2005-01-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service