Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670472

Sigma-Aldrich

1,2-Bis[(2R,5R)-2,5-dimethylphospholano]-3,3,4,4-tetrafluoro-1-cyclobutene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

95%

Synonym(s):

1,2-Bis[(2R,5R)-2,5-dimethyl-phospholanyl]3,3,4,4-tetrafluoro-1-cyclobutene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, catASium® MQF(R)Rh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36BF8P2Rh
CAS Number:
Molecular Weight:
652.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Quality Level

Assay

≥95% (31P-NMR)
95%

SMILES string

[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.C[C@@H]2CC[C@@H](C)P2C3=C(P4[C@H](C)CC[C@H]4C)C(F)(F)C3(F)F

InChI

1S/C16H24F4P2.C8H12.BF4.Rh/c1-9-5-6-10(2)21(9)13-14(16(19,20)15(13,17)18)22-11(3)7-8-12(22)4;1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h9-12H,5-8H2,1-4H3;1-2,7-8H,3-6H2;;/q;;-1;+1/b;2-1-,8-7-;;/t9-,10-,11-,12-;;;/m1.../s1

InChI key

FAEFUBUMCYELNL-HEEUKTGESA-N

General description

sold in collaboration with Solvias AG

Application

1,2-Bis[(2R,5R)-2,5-dimethylphospholano]-3,3,4,4-tetrafluoro-1-cyclobutene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate can be used as a catalyst in the hydrogenation of unsaturated lactate precursors and α-(hydroxymethyl)- acrylate derivatives.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

catASium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Chiral 2-Hydroxy-1-methylpropanoates by Rhodium-Catalyzed Stereoselective Hydrogenation of α-(Hydroxymethyl)-acrylate Derivatives
Holz J, et al.
advanced synthesis and catalysis, 350(16), 2533-2543 (2008)
Synthesis of Chiral 2-Hydroxy-1-methylpropanoates by Rhodium-Catalyzed Stereoselective Hydrogenation of α-(Hydroxymethyl)-acrylate Derivatives
Holz J, et al.
Advanced Synthesis & Catalysis, 350(16), 2533-2543 (2008)
Benjamin Schäffner et al.
ChemSusChem, 1(11), 934-940 (2008-10-29)
The asymmetric hydrogenation of alpha-acetoxy acrylates to O-acetyl lactates with Rh catalysts based on chiral bisphospholane ligands was investigated in propylene carbonate (PC) as "green" solvent. In contrast to DuPHOS-type ligands, catASium M ligands lead to full conversion of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service