Skip to Content
MilliporeSigma
All Photos(2)

Documents

67024

Sigma-Aldrich

(R)-1-Methyl hydrogen 3-methylglutarate

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(R)-Monomethyl 3-methylglutarate, (R)-mono-Methyl 3-methylglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
Beilstein:
1723141
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥98.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥90:10 (R:S) (GC)

refractive index

n20/D 1.438

density

1.116 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)C[C@H](C)CC(O)=O

InChI

1S/C7H12O4/c1-5(3-6(8)9)4-7(10)11-2/h5H,3-4H2,1-2H3,(H,8,9)/t5-/m1/s1

InChI key

BYBMHSADRRMVHY-RXMQYKEDSA-N

Other Notes

Chiral building block

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.F. Francis et al.
Journal of the Chemical Society. Chemical Communications, 579-579 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service