Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

669989

Sigma-Aldrich

(S)-α-Methoxy-2-naphthylacetic acid

97%

Synonym(s):

(S)-α-Methoxy-2-naphthaleneacetic acid, (S)-2-NMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)
97%

optical activity

[α]/D +177±5°, c = 0.1 in methanol

optical purity

enantiomeric excess: ≥99% (HPLC)

mp

111-114 °C

SMILES string

CO[C@H](C(O)=O)c1ccc2ccccc2c1

InChI

1S/C13H12O3/c1-16-12(13(14)15)11-7-6-9-4-2-3-5-10(9)8-11/h2-8,12H,1H3,(H,14,15)/t12-/m0/s1

InChI key

OQVFHDWREDKUCW-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service