Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668788

Sigma-Aldrich

S-Propyl chlorothioformate

96%

Synonym(s):

(Propylthio)carbonyl chloride, S-Propyl thiochloroformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SCOCl
CAS Number:
Molecular Weight:
138.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.4807

bp

37 °C/8 mmHg

density

1.149 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCSC(Cl)=O

InChI

1S/C4H7ClOS/c1-2-3-7-4(5)6/h2-3H2,1H3

InChI key

FYOPSYJKEHWOBR-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service