Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668176

Sigma-Aldrich

(−)-1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]benzene

kanata purity

Synonym(s):

(S,S)-i-Pr-DUPHOS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44P2
CAS Number:
Molecular Weight:
418.58
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

26-43 °C

SMILES string

CC(C)[C@@H]1CC[C@@H](C(C)C)P1c2ccccc2P3[C@@H](CC[C@H]3C(C)C)C(C)C

InChI

1S/C26H44P2/c1-17(2)21-13-14-22(18(3)4)27(21)25-11-9-10-12-26(25)28-23(19(5)6)15-16-24(28)20(7)8/h9-12,17-24H,13-16H2,1-8H3/t21-,22-,23-,24-/m0/s1

InChI key

RBVGOQHQBUPSGX-ZJZGAYNASA-N

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service