Skip to Content
MilliporeSigma
All Photos(1)

Documents

667900

Sigma-Aldrich

N-Boc-2-indolyldimethylsilanol

95%

Synonym(s):

2-(hydroxydimethylsilyl)-1,1-dimethylethylindole-1-carboxylic acid ester, N-Boc-indol-2-yldimethylsilanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((C8H5N)(CO2C(CH3)3))Si(CH3)2(OH)
CAS Number:
Molecular Weight:
291.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.545

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)n1c(cc2ccccc12)[Si](C)(C)O

InChI

1S/C15H21NO3Si/c1-15(2,3)19-14(17)16-12-9-7-6-8-11(12)10-13(16)20(4,5)18/h6-10,18H,1-5H3

InChI key

ARJIKJGACIMUOM-UHFFFAOYSA-N

Application

Reactant for:
Palladium-catalyzed cross-coupling reactions
A useful silicon nucleophile for Pd-catalyzed cross-coupling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Articles

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service