Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667633

Sigma-Aldrich

1-Benzyl-3-oxopiperazine

95%

Synonym(s):

4-(Phenylmethyl)-2-piperazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

148-155 °C

SMILES string

O=C1CN(CCN1)Cc2ccccc2

InChI

1S/C11H14N2O/c14-11-9-13(7-6-12-11)8-10-4-2-1-3-5-10/h1-5H,6-9H2,(H,12,14)

InChI key

SBWVHKNCFZRBRJ-UHFFFAOYSA-N

Application

Used in a copper-catalyzed amidation of an aryl bromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 43, 1101-1101 (2002)
Alessio Adamiano et al.
Analytical and bioanalytical chemistry, 406(24), 6021-6033 (2014-07-13)
Off-line analytical pyrolysis combined with gas chromatography–mass spectroscopy (GC–MS), directly or after trimethylsilylation, along with infrared spectroscopy and amino acid analysis was applied for the first time to the characterization of the intra-skeletal organic matrix (OM) extracted from four Mediterranean

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service